siRNA Details
Gene Symbol: 1700034P13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040462.1 | 2 | 1010 | |||
GenBank mRNA | AK006607.1 | - | 695 | |||
AK159293.1 | - | 1009 | ||||
AW106999.1 | - | 2 | 94 | |||
BB615872.1 | - | 2 | 178 |
