siRNA Details
Gene Symbol: 4930455B14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045968.1 | 6 | 892 | |||
GenBank mRNA | AK006409.1 | - | 4 | 678 | ||
AK015458.1 | - | 6 | 837 | |||
AK132658.1 | - | 5 | 783 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706466.1 | - | 4 | 678 | Chr. 14 - 8666390 - 8673320 [+] on Build GRCm38 |
