siRNA Details
Gene Symbol: Gm16894 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037980.1 | 4 | 521 | |||
GenBank mRNA | AK041998.1 | - | 4 | 524 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB638745.1 | - | 4 | 524 | Chr. 1 - 40076394 - 40085853 [-] on Build GRCm38 |
