siRNA Details
Gene Symbol: 1700039M10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006644.1 | - | 2 | 357 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706633.1 | - | 2 | 357 | Chr. 14 - 57571624 - 57572368 [+] on Build GRCm38 |
