siRNA Details
Gene Symbol: Gm26994 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK082412.1 | - | 5 | 561 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB650108.1 | - | 5 | 561 | Chr. 8 - 112556078 - 112569575 [-] on Build GRCm38 |
