siRNA Details
Gene Symbol: A330093E20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040342.1 | 4 | 663 | |||
GenBank mRNA | AK039712.1 | - | 4 | 583 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CK332073.1 | - | 3 | 473 | Chr. 18 - 45685130 - 45854544 [-] on Build GRCm38 |
