siRNA Details
Gene Symbol: 2310009A05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK003016.1 | - | 2 | 377 | ||
AK009244.1 | - | 2 | 384 | |||
BB027756.1 | - | 1 | 188 | |||
BC037369.1 | - | 2 | 351 | |||
BC100521.1 | - | 2 | 376 |
