siRNA Details
Gene Symbol: Gm10637 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK139810.1 | - | 3 | 544 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY725798.1 | - | 3 | 541 | Chr. 8 - 87197672 - 87199847 [-] on Build GRCm38 |
