siRNA Details
Gene Symbol: 5730419F03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK017575.1 | - | 262 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY717356.1 | - | 1, 2 | 259 | Chr. 1 - 78596263 - 78609506 [-] on Build GRCm38 |
