siRNA Details
Gene Symbol: 1700066N21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045924.1 | 5 | 725 | |||
GenBank mRNA | AK006908.1 | - | 5 | 726 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 1700066N21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045924.1 | 5 | 725 | |||
GenBank mRNA | AK006908.1 | - | 5 | 726 |