siRNA Details
Gene Symbol: 1700066N21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045924.1 | 1 | 295 | |||
GenBank mRNA | AK006908.1 | - | 1 | 296 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706904.1 | - | 1 | 300 | Chr. 5 - 87908583 - 87979351 [-] on Build GRCm38 |
