siRNA Details
Gene Symbol: 1700025K24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045825.1 | 3 | 707 | |||
GenBank mRNA | AK006354.1 | - | 3 | 709 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 1700025K24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045825.1 | 3 | 707 | |||
GenBank mRNA | AK006354.1 | - | 3 | 709 |