siRNA Details
Gene Symbol: Gm28515 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK136272.1 | - | 1 | 1996 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB064740.2 | - | 1 | 163 | Chr. 8 - 91907565 - 91911235 [-] on Build GRCm38 |
