siRNA Details
Gene Symbol: 1700074H08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045296.1 | 3 | 522 | |||
GenBank mRNA | AK006947.1 | - | 3 | 533 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BU936610.1 | - | 510 | Chr. - - [-] on Build |
