siRNA Details
Gene Symbol: Pinc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_003202.1 | 3 | 325 | |||
GenBank mRNA | DQ059755.1 | - | LIT3460 PINC |
3 | 325 | |
DQ059756.1 | - | LIT3461 PINC |
4 | 489 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BF124510.1 | - | 3 | 305 | Chr. 1 - 73392320 - 73407551 [-] on Build GRCm38 |
