siRNA Details
Gene Symbol: 1700023F02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038039.1 | 3 | 323 | |||
GenBank mRNA | AK006272.1 | - | 3 | 324 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706353.1 | - | 3 | 324 | Chr. 10 - 66120611 - 66124064 [-] on Build GRCm38 |
