siRNA Details
Gene Symbol: Gm28856 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK087224.1 | - | 159 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB664370.1 | - | 1 | 154 | Chr. 1 - 130766388 - 130782240 [+] on Build GRCm38 |
