siRNA Details
Gene Symbol: Gpr55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK131861.1 | - | 2 | 163 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY708138.1 | - | 2 | 163 | Chr. 1 - 85972365 - 85986917 [-] on Build GRCm38 |
