siRNA Details
Gene Symbol: 1700047M11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK047804.1 | - | 1938 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB360438.2 | - | 1 | 208 | Chr. 1 - 182302573 - 182303247 [+] on Build GRCm38 |
