siRNA Details
Gene Symbol: 1700030O20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045345.1 | 2, 3 | 409 | |||
GenBank mRNA | AK006570.1 | - | 2, 3 | 410 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706579.1 | - | 2, 3 | 405 | Chr. 10 - 116723067 - 116729173 [-] on Build GRCm38 |
