siRNA Details
Gene Symbol: 1810053B23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040486.1 | 2 | 269 | |||
NR_040487.1 | 3 | 264 | ||||
NR_040488.1 | 2 | 111 | ||||
NR_040489.1 | 2 | 109 | ||||
NR_040490.1 | 2 | 111 | ||||
NR_040491.1 | 3 | 189 | ||||
GenBank mRNA | AI390631.1 | - | 2 | 101 | ||
AK007854.1 | - | 2 | 270 | |||
AK019036.1 | - | 2 | 112 | |||
AK160158.1 | - | 2 | 110 | |||
AK160437.1 | - | 3 | 265 | |||
BM055834.1 | - | 3 | 189 | |||
BY707451.1 | - | 3 | 265 |
