siRNA Details
Gene Symbol: 1700041C23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033784.1 | 5 | 314 | |||
GenBank mRNA | AK006669.1 | - | 5 | 316 | ||
AK007035.1 | - | 5 | 315 | |||
AK018934.1 | - | 5 | 304 | |||
BY707168.1 | - | 5 | 304 |
