siRNA Details
Gene Symbol: 1700044K03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033785.1 | 2 | 223 | |||
GenBank mRNA | AK006696.1 | - | 2 | 224 | ||
AK018859.1 | - | 2 | 224 | |||
AK018880.1 | - | 2 | 224 | |||
BY706767.1 | - | 2 | 224 |
