siRNA Details
Gene Symbol: Gm5833 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK171408.1 | - | 3 | 270 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY750077.1 | - | 3 | 270 | Chr. 1 - 138671434 - 138682067 [-] on Build GRCm38 |
