siRNA Details
Gene Symbol: LOC102636514 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110474.1 | 1, 2 | 176 |
Gene Symbol: AV026068 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BU581897.1 | - | 1, 2 | 225 | ||
BX513262.1 | - | 1, 2 | 84 |
