siRNA Details
Gene Symbol: 2310039L15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045337.1 | 2 | 317 | |||
GenBank mRNA | AK009705.1 | - | 2 | 318 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY709350.1 | - | 2 | 318 | Chr. 10 - 95336276 - 95363216 [+] on Build GRCm38 |
