siRNA Details
Gene Symbol: 1700120C14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045627.2 | 1 | 35 | |||
GenBank mRNA | AK007210.1 | - | 1 | 36 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY707301.1 | - | 1 | 36 | Chr. 15 - 99251788 - 99262042 [+] on Build GRCm38 |
