siRNA Details
Gene Symbol: LOC100043315 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015482.1 | 3 | 535 | |||
GenBank mRNA | BC106998.1 | - | 3 | 626 | ||
BC106999.1 | - | 3 | 535 | |||
BC114343.1 | - | 3 | 535 | |||
BC115945.1 | - | 3 | 535 | |||
BC115946.1 | - | 3 | 535 | |||
BC127134.1 | - | 3 | 535 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY707425.1 | - | 4 | 851 | Chr. 15 - 95097961 - 95162394 [-] on Build GRCm38 |
