siRNA Details
Gene Symbol: 1700003P14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045982.1 | 4 | 902 | |||
GenBank mRNA | AK005657.1 | - | 4 | 903 | ||
AK016350.1 | - | 912 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 1700003P14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045982.1 | 4 | 902 | |||
GenBank mRNA | AK005657.1 | - | 4 | 903 | ||
AK016350.1 | - | 912 |