siRNA Details
Gene Symbol: 1700063D05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040392.1 | 4 | 519 | |||
GenBank mRNA | AK018871.1 | - | 4 | 516 | ||
BC048584.1 | - | 4 | 486 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BU938569.1 | - | 4 | 489 | Chr. 9 - 41206186 - 41217571 [+] on Build GRCm38 |
