siRNA Details
Gene Symbol: 2010009K17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040609.1 | 1 | 70 | |||
GenBank mRNA | AK008166.1 | - | 1 | 74 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY708124.1 | - | 1 | 73 | Chr. 2 - 158070364 - 158091797 [-] on Build GRCm38 |
