siRNA Details
Gene Symbol: 1700030N03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045304.1 | 2 | 122 | |||
GenBank mRNA | AK006563.1 | - | 118 | |||
AK007025.1 | - | 2 | 123 |
Gene Symbol: Gm38527 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK077035.1 | - | 134 |
