siRNA Details
Gene Symbol: 1700008K24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038141.1 | 2 | 386 | |||
NR_038142.1 | 3 | 289 | ||||
NR_038143.1 | 3 | 295 | ||||
NR_038144.1 | 2 | 242 | ||||
NR_038145.1 | 2 | 125 | ||||
GenBank mRNA | AK005780.1 | - | 3 | 273 | ||
CF104157.1 | - | 2 | 252 | |||
CF982619.1 | - | 3 | 236 |
