siRNA Details
Gene Symbol: 1700003L19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040507.1 | 5 | 667 | |||
GenBank mRNA | AK005649.1 | - | 5 | 668 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY705840.1 | - | 5 | 668 | Chr. 16 - 12811390 - 12848587 [+] on Build GRCm38 |
