siRNA Details
Gene Symbol: Gm19461 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037984.1 | 3 | 1034 | |||
GenBank mRNA | AK035267.1 | - | 2 | 573 | ||
AK145464.1 | - | 3 | 782 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB627542.1 | - | 2 | 570 | Chr. 1 - 133256546 - 133258475 [-] on Build GRCm38 |
