siRNA Details
Gene Symbol: 1700094J05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040580.1 | 3 | 598 | |||
GenBank mRNA | AK007067.1 | - | 3 | 597 | ||
AK019782.1 | - | 1 | 140 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715828.1 | - | 1 | 140 | Chr. 10 - 76398593 - 76398968 [+] on Build GRCm38 |
