siRNA Details
Gene Symbol: 4933400C23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040770.1 | 1 | 277 | |||
GenBank mRNA | AK077069.1 | - | 1 | 278 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB573726.2 | - | 1 | 278 | Chr. 9 - 92718716 - 92794629 [+] on Build GRCm38 |
