siRNA Details
Gene Symbol: 1700047L14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040691.1 | 3 | 430 | |||
GenBank mRNA | AK006716.1 | - | 3 | 431 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 1700047L14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040691.1 | 3 | 430 | |||
GenBank mRNA | AK006716.1 | - | 3 | 431 |