siRNA Details
Gene Symbol: 2210420H20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045389.1 | 1 | 210 | |||
GenBank mRNA | AK008989.1 | - | 1 | 212 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY708873.1 | - | 1 | 212 | Chr. 18 - 82754848 - 82759041 [-] on Build GRCm38 |
