siRNA Details
Gene Symbol: 2210420H20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045389.1 | 3, 4 | 570 | |||
GenBank mRNA | AK008989.1 | - | 3, 4 | 572 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 2210420H20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045389.1 | 3, 4 | 570 | |||
GenBank mRNA | AK008989.1 | - | 3, 4 | 572 |