siRNA Details
Gene Symbol: 1700006H21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045900.2 | 3 | 551 | |||
GenBank mRNA | AK005677.1 | - | 3 | 552 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY705860.1 | - | 3 | 552 | Chr. 13 - 107687397 - 107692169 [+] on Build GRCm38 |
