siRNA Details
Gene Symbol: 1700126H18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040695.1 | 2, 3 | 251 | |||
GenBank mRNA | AK007290.1 | - | 2, 3 | 252 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY707392.1 | - | 2, 3 | 252 | Chr. 5 - 66166607 - 66191335 [-] on Build GRCm38 |
