siRNA Details
Gene Symbol: C2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104191.1 | 3 | 685 | |||
GenBank mRNA | BX100594.1 | - | 3 | 684 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000630806.1 | - | 685 | Chr. - - [-] on Build |
