siRNA Details
Gene Symbol: LINC01163 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120619.1 | 6 | 2642 | |||
GenBank mRNA | AK124226.1 | - | 7 | 2716 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000630252.1 | - | 2717 | Chr. - - [-] on Build |
