siRNA Details
Gene Symbol: UBXN6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017027325.1 | XP_016882814.1 | 1 | 181 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000591414.5 | - | 1 | 181 | Chr. 19 - 4457962 - 4458759 [-] on Build GRCh38 |
