siRNA Details
Gene Symbol: LINC00311 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038859.1 | 2 | 858 | |||
GenBank mRNA | BC030801.1 | - | 2 | 858 | 4399117 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000366314.4 | - | 1 | 858 | Chr. 17 - 17858246 - 17860041 [+] on Build GRCh38 |
