siRNA Details
Gene Symbol: LOC101929130 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135551.1 | 2 | 778 | |||
GenBank mRNA | BX648496.1 | - | 2 | 779 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000636210.1 | - | 778 | Chr. - - [-] on Build |
