siRNA Details
Gene Symbol: SNHG25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_132278.1 | 2 | 101 | |||
GenBank mRNA | BM850443.1 | - | 2 | 101 | ||
CA337426.1 | - | 102 | ||||
CD050382.1 | - | 118 | ||||
CD388219.1 | - | 111 |
