siRNA Details
Gene Symbol: LINC01620 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_132343.1 | 2, 3 | 242 | |||
GenBank mRNA | BX114953.1 | - | 2, 3 | 226 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000306731.4 | - | 2, 3 | 226 | Chr. 20 - 44451984 - 44465328 [-] on Build GRCh38 |
