siRNA Details
Gene Symbol: LINC01620 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_132342.1 | 2 | 625 | |||
GenBank mRNA | BC030259.1 | - | 2 | 626 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000623295.1 | - | 625 | Chr. - - [-] on Build |
